VWR® Round Media Bottles
Supplier: VWR International
89166-092 89166-094 89166-096 89166-098 89166-100 89166-102 89166-104 89166-106 89166-108
89166-092CS 764.59 CAD
89166-092 89166-094 89166-096 89166-098 89166-100 89166-102 89166-104 89166-106 89166-108
VWR® Round Media Bottles
Bottles Cell Culture Bottles
Disposable round media bottles for storing sterile solutions such as tissue culture media, serum, and buffers.
  • Polycarbonate or polystyrene construction
  • Sloped bottle design
  • Easy to grip
  • Easily visible graduations
Bottles feature a sloped design that improves stability on the work surface.
Polycarbonate bottles can withstand up to three autoclave cycles and offer good alcohol resistance. They are stable up to 135 °C (275 °F). Polystyrene bottles are ideal for general-purpose cell culture aqueous solutions, and are stable up to 80 °C (176 °F). PETGMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable