Microflex® XCEED® XC-310 Nitrile Disposable Gloves
Supplier: Microflex
XC-310-XS XC-310-S XC-310-M XC-310-L XC-310-XL
CA89174-538BX 71.18 CAD
CA89174-538 CA89174-540 CA89174-542 CA89174-544 CA89174-546
Microflex® XCEED® XC-310 Nitrile Disposable Gloves
Gloves
Ergonomically designed to reduce hand fatigue while maintaining excellent tactile sensitivity.
  • Made without accelerators to protect from allergic reactions
  • ERGOFORM™ Technology; an ergonomic design that supports musculoskeletal health
  • Gloves offer advanced barrier integrity with a low 0.65 AQL
  • Fentanyl-tested for enhanced user protection
Microflex® XCEED® XC-310 gloves are manufactured without accelerators for low dermatitis potential. These gloves prevent instances of Type I latex allergies and Type IV chemical allergic reactions. To ensure extra durabilit and enhanced comfort, the stronger yet softer formulation protects against rips and tears while keeping the same tactile sensitivity and a lightweight, softer skin feel. These gloves are designed with the proprietary ERGOFORM™ Technology to support musculoskeletal health and increase worker productivity.
Gloves are smartly packaged, with 250 gloves per box and easy, single glove dispensing to reduce waste. These medical exam gloves are specially tested against fentanyl to ensure advanced protection and peace of mind.
Certifications: ASTM D6319, ASTM D6978, Food Type: Rubber, EN 1149-1/2/3, EN 420:2003 + A1:2009, FDA21 CFR 177-2600-US Food Contact Approved, US QSR/FDA 510(k) Medical Examination Grade.
Packaging: Gloves are packaged 250 pieces per dispenser, 10 dispensers per case.