SP Bel-Art Bench-Top Biohazard Bags, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a part of SP
SP Bel-Art
F13166-0001 F13166-0000
70690-104PK 579.12 CAD
70690-104 11215-857
SP Bel-Art Bench-Top Biohazard Bags, Bel-Art Products, a part of SP
Bags Waste Disposal Bags
Bright red, autoclavable bags feature one long flap for easy opening. Printed with the biohazard symbol and sterilization indicator patch, they are made of High Molecular Weight, High-Density (HMHD) polyethylene.
  • Use as a biohazard material handler: Small size allows the bag to be inverted over your hand to safely pick up waste
  • Steam autoclavable at 121 °C (250 °F)
For a space-saving disposal unit in fume hoods or on bench tops, use with Poxygrid® Benchtop Biohazard Bag Holder (11215-508) and Cover (89237-571).
Certifications: Pass ASTM 1922 tear resistance and ASTM 1709 dart impact tests.