SlimLine™ Stainless Steel Cart Systems, Micronova
Supplier: Micronova
C-22S C-33S C3A-03 C-227
68400-726EA 4906.07 CAD
68400-726 68400-728 94023-746 89423-426
SlimLine™ Stainless Steel Cart Systems, Micronova
Mops
Durable cart systems feature a stainless steel frame with push bar and three grip-it holders for easy storage and interchange of handles and additional mop heads. Systems come standard with color-coded, 22.7 liter (6 gal.) polypropylene buckets. \
  • Crafted from 304 electropolished stainless steel for durability and long life
  • Angled push bar for easy handling in the cleanroom
  • Color-coded, 22 L (6 gallon) polypropylene, or 30 L (8 gallon) stainlesss steel buckets included
  • Optional wringer bar allows wringers to be placed above the rinse bucket
  • Prevenz™ antimicrobial wheels
  • Foot activated brake, entire unit is autoclavable
  • Buckets are best autoclaved in the inverted position
The Micronova SlimLine™ Stainless Steel Cart Systems with either polypropylene plastic or stainless steel buckets, accommodate a range of mop wringers. An optional wringer bar, designed for use with the W-6 Downpress Wringer, allows placement above the rinse or empty third bucket for simplified wringing.
Model 89423-426 includes two 30 liter (8 gal.) buckets. The 68400-726 and 68400-728 carts inlcude the 6 gallon plastic buckets. The 89423-426 includes two 8 gallon buckets, made from molded stainless steel with no seams or ridges to harbor bacteria.
Accessories information: Buckets are included with carts.
Ordering information: Replacement buckets and replacement wheels (89413-040) are also available. All other components must be ordered separately.