Automatic Switchover Systems, Ceodeux
Supplier: Ceodeux Inc.
TDL200125580 TDL200125540 TDI200125320 TDI200125350
89133-262EA 6875.15 CAD
89133-262 89133-266 89133-272 89133-276
Automatic Switchover Systems, Ceodeux
Gas Regulators
Switchover systems are designed to minimize downtime by ensuring a constant flow of gas to instrument
All systems are equipped with two flexible stainless steel pigtails with CGA connections. Systems also feature integral check valves in CGA inlet glands, an SL10 line regulator, and 6mm (1/4") NPT outlets.
The system automatically recognizes and alternates service from a primary cylinder to a reserve cylinder as gas is depleted. Chrome-plated brass systems are suitable for the supply of high-purity, non-corrosive gases in low flow applications. Stainless steel systems are suitable forMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable