Puritan® PurCollect Environmental Large Foam Tipped Sampling Swab, Polypropylene Handle with Thumb Stop, Sterile, Puritan Medical Products
Supplier: Puritan Medical Products
25-16071PFSC 25-16071PFSC
CA89087-866CS 1873.5 CAD
CA89087-866 89087-866
Puritan® PurCollect Environmental Large Foam Tipped Sampling Swab, Polypropylene Handle with Thumb Stop, Sterile, Puritan Medical Products
Swabs Swabs for Sampling
Environmental sampling device features a polypropylene shaft with prominent thumb stop to indicate hand placement
Made in the USA.
Shaft is scored 40mm (19/16") from end of tip to allow easy detachment. Large macrofoam tip for maximum surface collection.
Certifications: CE marked.
Ordering information: Packaged as 10 boxes of 50 individually wrapped swabs.