Puritan® Large Paddle Shaped Knitted Polyester Tipped Applicator, Solid Polypropylene Handle, Puritan Medical Products
Supplier: Puritan Medical Products
3655 3655
CA89133-800CS 2116.5 CAD
CA89133-800 89133-800
Puritan® Large Paddle Shaped Knitted Polyester Tipped Applicator, Solid Polypropylene Handle, Puritan Medical Products
Swabs Swabs for Sampling
Used in manufacturing or refurshing of electronic and communcation devices as well as removing residues and particulates
Solid polypropylene shaft. Large paddle tip. Molded handle for easy grip. Low in NVR's and particulates. Lint free. Nonsterile. Made in the USA.
Certifications: CE marked.
Ordering information: Packaged 100 per pack, 10 packs per case.