PurSwab® Small Mitt Soft Foam Tipped Applicator, Polypropylene Handle, Puritan Medical Products
Supplier: Puritan Medical Products
1135 1135
CA89133-792CS 1148.25 CAD
CA89133-792 89133-792
PurSwab® Small Mitt Soft Foam Tipped Applicator, Polypropylene Handle, Puritan Medical Products
Swabs Swabs for Sampling
For precision cleaning
Made in the USA.
Rigid pointed polypropylene handle under soft oval foam mitt. Good absorbency. Can be used with IPA and DIA water. Lint and residue free. Nonsterile.
Certifications: CE marked.
Ordering information: Packaged 100 per pack, 10 packs per case.