Pur-Wraps® Swab, Cotton Tip, Sterile or Nonsterile, Puritan Medical Products
Supplier: Puritan Medical Products
803-PC 25-8061PC 803-PC 25-8062PC 806-PC 25-8061PC
CA10805-140CS 1062.76 CAD
CA10805-140 CA10805-144 CA10805-148 CA10806-050 10805-140 10805-144 10805-148 10806-050
Pur-Wraps® Swab, Cotton Tip, Sterile or Nonsterile, Puritan Medical Products
Swabs Swabs for Sampling
This absorbent cotton tipped applicator is produced with high-quality USP grade cotton, which is carded and coiled onsite to maintain consistency.
It is bonded securely to a semi-flexible polystyrene handle by an aqueous-based adhesive. Available either nonsterile or sterile.