Puritan® Polyester Tip Applicator, Wood Handle, Sterile, Puritan Medical Products
Supplier: Puritan Medical Products
25-8061WD 25-8061WD 25-8062WD
CA10805-160CS 1605.15 CAD
CA10805-160 CA89133-768 10805-160 89133-768
Puritan® Polyester Tip Applicator, Wood Handle, Sterile, Puritan Medical Products
Swabs Swabs for Sampling
Regular, non-absorbent polyester tip is bonded to a sturdy wooden handle with an aqueous adhesive
It is ideal for specimen collection, patient care, and surface collection. Made in the USA.
Certifications: CE marked.