PurSwab® Mini-Tip Foam Tipped Applicator, Polypropylene Handle, Puritan Medical Products
Supplier: Puritan Medical Products
PurSwab®
1803-PFB 1803-PFB
10805-138CS 603.76 CAD
10805-138 CA10805-138
PurSwab® Mini-Tip Foam Tipped Applicator, Polypropylene Handle, Puritan Medical Products
Swabs Swabs for Sampling
For removing epoxy residue from connectors and general cleaning
Made in the USA.
Black polypropylene shaft. Flexible mini foam tip. Can be used with IPA and DI water or dry. Lint and residue free. Nonsterile.
Certifications: CE marked.
Ordering information: Packaged as 2 bags of 500 swabs per bag.