PharmaMOP® Mop System, Foamtec International
Supplier: Foamtec International
FS853ST FS856ST FS853ENST
89107-502CS 2084.95 CAD
89107-502 89107-506 89133-232
PharmaMOP® Mop System, Foamtec International
Mops
Designed for all critical cleanroom surfaces, PharmaMOP® Mop System allows cleaning of ceilings, walls, and floors with ease.
  • Mop heads feature polyester over foam construction
  • Abrasion-free mopping for ISO Class 1–7 cleanrooms
  • Gamma irradiated to SAL 10¯⁶
System combines a unique mop head optimized to meet label-indicated disinfectant contact times with an ultra-lightweight, telescoping handle and frame for unsurpassed cleaning efficiency
Mop frame features a two-direction 360° swivel and 3.8cm (1.5") low-profile design that enables unsurpassed access to tight spaces, reducing uncleanable dead spots. Adjustable to lengths of 127cm (50"), 144.8cm (57"), and 162.6cm (64").
Enhances SOP cleaning compliance through improved ergonomics. Utilizes a simple press fit connection that enables gloved operators to easily attach mop heads. Compatible with steam sterilization processes.
Certifications: Mop heads are lot certified, traceable back to raw materials, and autoclave compatible.
Ordering information: Mop head and frame are sold separately. Mop heads and components are individually packed. Standard mop heads, extension shafts, and replacement press clips are available. Contact your Avantor representative for more information.