VWR® Sterile Sleeves
Supplier: VWR International
414004-510 414004-511 76169-446 76169-448
414004-510CS 1002.31 CAD
414004-510 414004-511 76169-446 76169-448
VWR® Sterile Sleeves
Disposable Garments
VWR® Sterile Sleeves are manufactured from a specially formulated breathable microporous fabric that provides significant fluid and barrier protection. Garments exhibit excellent water vapor transmission to optimize user comfort.
  • Excellent breathability and water vapor transmission rate
  • Superior fluid and particulate barrier
  • Lowest level of particle shedding
  • Durable yet soft, cloth-like fabric
  • Gamma irradiated to SAL of 10-6
Sleeves feature elastic openings and sonically welded seams. They are available in two versions; with thumb loops or without.