TOC Cleaning Validation Kits, Texwipe®
Supplier: Texwipe
TX3342 TX3340
TWTX3342EA 1184.57 CAD
TWTX3342 TWTX3340
TOC Cleaning Validation Kits, Texwipe®
Swabs Swabs for Cleaning
Texwipe’s Cleaning Validation Kits were designed to simplify sampling as part of a cleaning validation protocol. The swabs and vials are further cleaned using a proprietary process to make these kits excellent for use in Total Organic Carbon (TOC) analysis.
  • Swab head is cleanroom laundered, double-layered polyester knit
  • Break-away handle allows the swab head to be placed into a sampling vial with minimal handling
Kits contain vials, swabs, and labels to sample multiple places in the production area. Prewashed TOC vials have been manufactured, analyzed,More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable