Puritan® Thin Paddle Shaped Knitted Polyester Tipped Applicator, Polypropylene Handle, Puritan Medical Products
Supplier: Puritan Medical Products
3140 3140
CA89133-802CS 1804.5 CAD
CA89133-802 89133-802
Puritan® Thin Paddle Shaped Knitted Polyester Tipped Applicator, Polypropylene Handle, Puritan Medical Products
Swabs Swabs for Sampling
Cleaning swab is used in manufacturing, or refurbishing of electronic and communication devices as well as removing residues and particulates
Polypropylene shaft. Flexible thin paddle tip. Low in NVR’s and particulates. Chemical resistant. Lint free. Nonsterile. Made in the USA.
Certifications: CE marked.
Ordering information: Packaged 100 per pack, 10 packs per case.