DuPont™ Tyvek® IsoClean® Bouffant Caps
Supplier: DuPont
IC729SWH0002500B IC729SWH000250CS IC729SWH0002500B
CA89125-652CS 2061.46 CAD
CA89125-652 CA89127-360 89125-652
DuPont™ Tyvek® IsoClean® Bouffant Caps
Disposable Garments
Tyvek® IsoClean® delivers an ideal balance of protection, durability and comfort.
  • Serged seams
  • Elastic headband
  • One size fits most
Made using a patented flash spinning process, Tyvek® provides an inherent barrier to particles, microorganisms and non-hazardous light liquid splash. Serged seams have multiple interlocking threads that are sewn around the raw edges of the garment material to create a strong, stress-resistance seam. Diameter of head opening is 211/2" stretched and 6" relaxed. Elastic headband allows for one size to fit most.
Clean-processed garments are processed to minimize particle shedding.
Certifications: Sterile garments are gamma sterilized to SAL of 10-6.
Packaging: Bulk packaged option packed in double transparent poly liners.