Polyethylene Bag Systems, Sterile, Advanced Scientifics
Supplier: Advanced Scientific, Inc.
SS00040-I SS00041-I SS00103-I SS00104-I
89127-080CS 4107.45 CAD
89127-080 89127-082 89127-130 89127-132 89127-134
Polyethylene Bag Systems, Sterile, Advanced Scientifics
Bags Media Bags
Polyethylene single-use bag systems are produced from two films.
The fluid contact film is 5 mil polyethylene. The outer layer is five-layer 7 mil coextrusion film. Manufactured in an ISO Class 7 (FED-STD-209E Class 10000/M5.5) cleanroom environment. Sterile.
Certifications: ISO 13485 certified. Complies with cGMP manufacturing regulations. FDA registered.