Ethyl Vinyl Acetate (EVA) Bag Systems, Sterile, Advanced Scientifics
Supplier: Advanced Scientific, Inc.
SS00001-I SS00002-I SS00003-I SS00006-I SS00007-I SS00008-I SS00009-I SS00010-I SS00011-I SS00012-I SS00013-I SS00016-I SS00017-I SS00018-I SS00019-I SS00021-I SS00022-I SS00023-I
89127-032CS 3932.41 CAD
89127-032 89127-034 89127-036 89127-042 89127-044 89127-046 89127-048 89127-050 89127-052 89127-054 89127-056 89127-058 89127-062 89127-064 89127-066 89127-068 89127-070 89127-072 89127-074 89127-076
Ethyl Vinyl Acetate (EVA) Bag Systems, Sterile, Advanced Scientifics
Bags Media Bags
Ethyl vinyl acetate (EVA) single-use bag systems are produced from a Four-layer 12.5 mil coextrusion film for a durable barrier.
  • Irradiated and sterile
  • Eliminating the concern of cross-contamination
Manufactured in an ISO Class 7 (FED-STF-209E Class 10,000/M5.5) cleanroom environment. Ideal for biopharmaceutical, laboratory, and diagnostic applications.
Certifications: EVA film meets United States Pharmacopoeia Class VI Plastics Guidelines. ISO 13485 certified. Complies with cGMP manufacturing regulations. FDA registered.