DuPont™ Tyvek® IsoClean® Sleeves
Supplier: DuPont
IC501BWH0001000S IC501BWH0001000B IC501BWH0001000C IC501BWH000100CS IC501BWH0001000S IC501BWH0001000C IC501BWH000100CS
CA89125-646CS 998.02 CAD
CA89125-646 CA89127-350 CA89127-352 CA89127-354 89125-646 89127-352 89127-354
DuPont™ Tyvek® IsoClean® Sleeves
Disposable Garments
Tyvek® IsoClean® delivers an ideal balance of protection, durability and comfort.
  • Elastic at both ends
  • One size fits most
  • Serged seams
Made using a patented flash spinning process, Tyvek® provides an inherent barrier to particles, microorganisms and non-hazardous light liquid splash. Serged seams have multiple interlocking threads that are sewn around the raw edges of garment material to create a strong, stress-resistant seam. Elastic at both ends allows for a secure fit.
Clean-processed garments offer lowest level of particle shedding within DuPont product portfolio.
Certifications: Sterile garments are gamma sterilized to SAL of 10-6.
Packaging: Bulk packaged option packed in double transparent poly liners.