DuPont™ Tyvek® IsoClean® Shoe Covers with PVC Sole
Supplier: DuPont
IC461SWHMD03000B IC461SWHSM03000B IC461SWHXL03000B IC461SWHLG03000B
CA89127-344CS 2550.14 CAD
CA89127-344 CA89127-346 CA89127-348 CA89125-644
DuPont™ Tyvek® IsoClean® Shoe Covers with PVC Sole
Disposable Garments
Tyvek® IsoClean® delivers an ideal balance of protection, durability and comfort.
  • PVC sole
  • Elastic opening
  • Serged seams
Made using a flash spinning process, Tyvek® provides an inherent barrier to particles, microorganisms and non-hazardous light liquid splash.
Serged seams have multiple interlocking threads that are sewn around the raw edges of garment material to create a strong, stress-resistant seam. Elastic at ankles allows for a secure fit. PVC soles provide enhanced skid-resistance and durability.
Packaging: Bulk packaged garments are bulk packaged in double transparent poly liners.