DuPont™ Tyvek® IsoClean® Shoe Covers with Gripper™ Sole
Supplier: DuPont
IC451SWHLG01000B IC451SWHMD01000B
CA89127-320CS 678.26 CAD
CA89127-320 CA89127-322 CA89129-682
DuPont™ Tyvek® IsoClean® Shoe Covers with Gripper™ Sole
Disposable Garments
Tyvek® IsoClean® delivers an ideal balance of protection, durability and comfort.
  • Gripper™ soles provide enhanced skid-resistance and durability
  • Enhanced skid-resistance and durability
  • Elastic at ankles allows for a secure fit
Made using a patented flash spinning process, Tyvek® provides an inherent barrier to particles, microorganisms and non-hazardous light liquid splash.
Serged seams have multiple interlocking threads that are sewn around the raw edges of garment material to create a strong, stress-resistant seam.
Packaging: Bulk packaged garments are bulk packaged in double transparent poly liners.