DuPont™ Tyvek® IsoClean® Boot Covers with PVC Sole
Supplier: DuPont
IC457SWHLG01000B IC457SWHSM01000S IC457SWHLG01000S IC457SWHMD01000S IC457SWHXL01000S IC457SWHSM01000B IC457SWHMD01000B IC457SWHLG01000B IC457SWHMD01000B IC457SWHXL01000S
CA89125-636CS 1539.93 CAD
CA89125-636 CA89125-638 CA89127-324 CA89127-328 CA89127-330 CA89128-796 CA89127-326 89125-636 89127-326 89127-332
DuPont™ Tyvek® IsoClean® Boot Covers with PVC Sole
Disposable Garments
Tyvek® IsoClean® delivers an ideal balance of protection, durability and comfort.
  • PVC sole
  • Elastic opening
  • Ankle ties
  • Coverage up to calf
  • Serged seams
Made using a patented flash spinning process, Tyvek® provides an inherent barrier to particles, microorganisms and non-hazardous light liquid splash.
Serged seams have multiple interlocking threads that are sewn around the raw edges of garment material to create a strong, stress-resistant seam. Ankle ties allow for a secure fit. PVC soles provide enhanced skid-resistance and durability.
Certifications: Sterile garments are gamma sterilized to SAL of 10-6.
Packaging: Bulk packaged option packed in double transparent poly liners. Clean Processed, Sterile garments are individually packaged.