DuPont™ Tyvek® IsoClean® Frocks with High Mandarin Collar and Set Sleeves
Supplier: DuPont
IC263SWH2X00300B IC263SWH3X00300B IC263SWH4X00300B IC263SWHSM00300B IC263SWHMD00300B IC263SWHLG00300B
CA89125-622CS 742.34 CAD
CA89125-622 CA89127-296 CA89127-298 CA89127-300 CA89127-304 89125-620 89127-302
DuPont™ Tyvek® IsoClean® Frocks with High Mandarin Collar and Set Sleeves
Disposable Garments
Tyvek® IsoClean® delivers an ideal balance of protection, durability and comfort.
  • Serged seams
  • High mandarin collar with adjustable snaps
  • Set sleeve design
  • Elastic wrists
  • Front snap closure
  • Generous sizing and cut
Made using a patented flash spinning process, Tyvek® provides an inherent barrier to particles, microorganisms and non-hazardous light liquid splash. Serged seams have multiple interlocking threads that are sewn around the raw edges of garment material to create a strong, stress-resistant seam. Elastic openings at wrists and high mandarinMore Product Information