DuPont™ Tyvek® IsoClean® Frocks with High Mandarin Collar and Raglan Sleeves
Supplier: DuPont
IC262SWH2X00300S IC262SWH3X00300B IC262SWH3X00300S IC262SWH4X00300B IC262SWHLG00300B IC262SWHLG00300S IC262SWHMD00300B IC262SWHMD00300S IC262SWHSM00300B IC262SWHXL00300B IC262SWHXL00300S IC262SWHLG00300B IC262SWHMD00300B IC262SWHXL00300B
CA89127-272CS 874.91 CAD
CA89127-272 CA89127-274 CA89127-276 CA89127-278 CA89127-280 CA89127-282 CA89127-284 CA89127-286 CA89127-288 CA89127-290 CA89127-292 CA89127-294 89127-282 89127-286 89127-292
DuPont™ Tyvek® IsoClean® Frocks with High Mandarin Collar and Raglan Sleeves
Disposable Garments
Tyvek® IsoClean® delivers an ideal balance of protection, durability and comfort.
  • Elastic openings at wrists and high mandarin collar with adjustable snaps allow for a secure fit
  • Raglan sleeve design allows for greater range of motion and comfort
  • Generous sizing and cut
Made using a flash spinning process, Tyvek® provides an inherent barrier to particles, microorganisms and non-hazardous light liquid splash. Serged seams have multiple interlocking threads that are sewn around the raw edges of garment material to create a strong, stress-resistant seam. Frocks provide body-length jacket design with front snap closures for easy donning and doffing.
Certifications: Sterile garments are gamma sterilized to SAL of 10-6.
Packaging: Bulk packaged garments are bulk packaged in double transparent poly liners.