DuPont™ Tyvek® IsoClean® Coveralls with Dolman Sleeves
Supplier: DuPont
IC253BWHLG0025CS IC253BWH2X00250C IC253BWH2X00250S IC253BWH2X0025CS IC253BWH3X00250C IC253BWH3X00250S IC253BWH3X0025CS IC253BWH4X00250S IC253BWH4X0025CS IC253BWH5X0025CS IC253BWHLG00250C IC253BWHLG00250S IC253BWHMD00250C IC253BWHMD00250S IC253BWHMD0025CS IC253BWHSM00250S IC253BWHSM0025CS IC253BWHXL00250C IC253BWHXL00250S IC253BWHXL0025CS IC253BWH2X00250B IC253BWH3X00250B IC253BWH4X00250B IC253BWH5X00250B IC253BWH5X00250S IC253BWHLG00250B IC253BWHMD00250B IC253BWHSM00250B IC253BWHSM00250C IC253BWHXL00250B IC253BWH2X0025CS IC253BWH3X0025CS
CA89125-616CS 4061.31 CAD
CA89125-616 CA89127-208 CA89127-210 CA89127-212 CA89127-214 CA89127-216 CA89127-218 CA89127-222 CA89127-224 CA89127-226 CA89127-228 CA89127-230 CA89127-232 CA89127-234 CA89127-236 CA89127-238 CA89127-240 CA89127-242 CA89127-244 CA89127-246 CA89129-640 CA89129-642 CA89129-644 CA89129-646 CA89129-650 CA89129-652 CA89129-654 CA89129-656 CA89129-658 CA89129-660 89127-212 89127-218
DuPont™ Tyvek® IsoClean® Coveralls with Dolman Sleeves
Disposable Garments
Tyvek® IsoClean® delivers an ideal balance of protection, durability and comfort.
  • Bound neck allows for lower particle shedding
  • Front zipper closure allows for easy donning and doffing
Made using a flash spinning process, Tyvek® provides an inherent barrier to particles, microorganisms and non-hazardous light liquid splash. Bound seams are covered with garment fabric to reinforce the seam and to reduce the potential for particle penetration. Coveralls feature elastic openings at wrists and ankles for a tighter fit. Dolman sleeve design allows for greater range of motion and comfort.
Clean-processed garments offer lowest level of particle shedding within DuPont product portfolio.
Certifications: Sterile garments are gamma sterilized to SAL of 10-6.
Packaging: Bulk packaged garments are bulk packaged in double transparent poly liners. Clean Processed and Clean Processed, Sterile garments are individually packaged.