DuPont™ Tyvek® IsoClean® Coveralls with Attached Hood, Boots, and Thumb Loops
Supplier: DuPont
IC105SWH2X0025CS IC105SWH3X0025CS IC105SWHLG0025CS IC105SWHMD0025CS IC105SWH2X00250C IC105SWH3X002500 IC105SWH3X00250C IC105SWHLG002500 IC105SWHLG00250C IC105SWHMD002500 IC105SWHMD00250C IC105SWHXL002500 IC105SWHXL00250C IC105SWHXL0025CS
CA89127-172CS 3973.29 CAD
CA89127-172 CA89127-174 CA89127-176 CA89127-178 CA89129-594 CA89129-596 CA89129-598 CA89129-600 CA89129-602 CA89129-604 CA89129-606 CA89129-608 CA89129-610 CA89129-612 CA89129-614
DuPont™ Tyvek® IsoClean® Coveralls with Attached Hood, Boots, and Thumb Loops
Disposable Garments
Tyvek® IsoClean® delivers an ideal balance of protection, durability and comfort.
  • Serged seams
  • Set sleeve design
  • Elastic wrists and ankles
  • Attached thumb loops
  • Zipper closure
  • Attached boots with PVC sole
Tyvek® provides an inherent barrier to particles, microorganisms and non-hazardous light liquid splash. Serged seams have multiple interlocking threads that are sewn around the raw edges of garment material to create a strong, stress-resistant seam. Coveralls have elastic openings for a tighter fit at wrists and ankles. Thumb loops minimize bunching sleeves. Attached hood with elastic around face opening ensures a secure fit. Front zipper closure allows for easy donning and doffing. Soles provide enhanced skid-resistance and durability.
Clean-processed garments offer lowest level of particle shedding within DuPont product portfolio.
Certifications: Sterile garments are gamma sterilized to SAL of 10-6.
Packaging: Bulk packaged garments are bulk packaged in double transparent poly liners. Clean Processed and Clean Processed, Sterile garments are individually packaged.