DuPont™ Tyvek® Micro-Clean® 2-1-2 Coveralls
Supplier: DuPont
Tyvek® Micro-Clean®
CC252BBU3XPI CC252BBU2X0S CC252BBULGPI CC252BBUSM0S CC252BBUSMPI CC252BBUXL0S CC252BBU2X0S CC252BBU2XPI CC252BBUMD0S CC252BBULG0S CC252BBU2XPI CC252BBUMD0S CC252BBU3X0S CC252BBUXL0S CC252BBUSMPI CC252BBU5X0S CC252BBU4X0S CC252BBU3XPI CC252BBU3X0S CC252BBUMDPI CC252BBUXLPI CC252BBULGPI CC252BBU4XPI CC252BBUMDPI CC252BBU4XPI CC252BBUSM0S CC252BBU4X0S CC252BBULG0S CC252BBU5X0S CC252BBUXLPI
CA89127-144CS 2246.87 CAD
CA89127-144 CA89127-166 89127-156 CA89127-164 89127-154 CA89127-148 CA89127-146 89127-158 89129-588 CA89127-162 CA89129-592 89127-152 CA89127-160 89127-150 CA89129-590 CA89129-588 89127-146 CA89127-154 89129-590 89127-144 89127-166 CA89127-152 CA89127-158 89127-148 CA89127-156 89127-160 89127-164 CA89127-150 89127-162 89129-592
DuPont™ Tyvek® Micro-Clean® 2-1-2 Coveralls
Disposable Garments
Tyvek® Micro-Clean® 2-1-2 delivers the particle barrier, durability and comfort of Tyvek® with a unique blue color.
  • Bound seams
  • Bound neck with loop at center back
  • Dolman sleeve design
  • Covered elastic wrists and ankles
  • Zipper closure
Proprietary blue polymeric resin coating is applied to Tyvek® fabric. Dolman sleeve design allows for greater range of motion and comfort. Coveralls have elastic openings for tighter fit at wrists and ankles. Covered elastic minimizes particle shedding. Bound seams are covered with garment fabric to reinforce the seam and to reduce the potential for particle penetration.
Certifications: Sterile garments are gamma sterilized to a Sterility Assurance Level (SAL) of 10-6.
Packaging: Coveralls are individually packaged in an opaque bag.