Tygon® A-60-F IB Temperature and Pressure-Resistant Food and Beverage Tubing
Supplier: Saint-Gobain
Tygon®
APW00019 APW00029
63008-830EA 929.15 CAD
63008-830 63008-832
Tygon® A-60-F IB Temperature and Pressure-Resistant Food and Beverage Tubing
Tubing Transfer Tubing
Versatile Tygon® A-60-F IB temperature and pressure-resistant food and beverage tubing provides long life for everyday use and consistently outperforms general-purpose rubber tubing in a wide variety of applications.
  • Temperature resistant from –75 to 275 °F
  • Ozone and UV light resistant
  • Repeatedly autoclavable, and compatible with a wide range of fluids
Tygon® A-60-F IB Pressure Tubing handles at least 4 times the pressure of non-reinforced tubing. It has excellent resistance to ozone and UV light, with little to no sign of deterioration in laboratory testing to the equivalent of 10 years outdoor exposure.
Tygon® Pressure Tubing is extremely abrasion resistant and bends easily to simplify custom component assembly. This tubing is ideal for use in clean-in-place and steam-in-place cleaning and sterilization systems. It is compatible with virtually all commercial cleaners and sanitizers, and can be repeatedly autoclaved up to five times without affecting the overall service life.
Tygon® A-60-F IB Pressure Tubing provides superior resistance to many corrosive fluids when compared to general-purpose rubber tubings. This tubing has excellent acid and alkali resistance, and is compatible with numerous oxidizing agents, such as hydrogen peroxide, sodium hypochlorite, and ozone. It also shows good resistance to the animal and vegetable oils frequently found in food contact applications.
Certifications: Meets FDA and 3-A Criteria.
Ordering information: Tygon® A-60-F IB Pressure Tubing is supplied in 50' lengths.