CONSTIX®, Sealed Foam Swabs, SF-1 Series, Contec®
Supplier: Contec
CONSTIX®
SF-1
68310-228CS 306.52 CAD
68310-228
CONSTIX®, Sealed Foam Swabs, SF-1 Series, Contec®
Swabs Swabs for Cleaning
CONSTIX® SF-1 sealed foam swabs consist of polyurethane foam bonded to a polypropylene handle without the use of adhesives or binders.
  • Polyurethane foam head provides good particle entrapment and removal
  • Large head is ideal for cleaning and applying coating
The absorbent foam enables solvents, such as isopropanol and ethanol, to be readily sorbed for applications involving solvent delivery or removal. Though compatible with many solvents, the SF-1 is not recommended for use with ketones, such as acetone or MEK.
Ordering information: Laundered and double bagged.