Sampling System, Sterile, Advanced Scientifics
Supplier: Advanced Scientific, Inc.
4MP0034 4MP0035 4MP0036 4MP0037 4MP0038 4MP0039
89127-372CS 2549.12 CAD
89127-372 89127-374 89127-376 89127-378 89127-380 89127-382
Sampling System, Sterile, Advanced Scientifics
Bags Sample Bags
Polyethylene sample bags complete with 30.5 cm (12") c-flex tubing, pinch and cut devices, male luer injection sites, and 3.8 cm (1½") sanitary sampling system valve.