Water, STERI-WATER® USP, purified, sterile

Supplier: VELTEK ASSOCIATES INC

Synonyms: Hydrogen oxide

STWA-02 STWA-5G STWA-2G STWA-16Z STWA-8L
76326-572CS 277.81 CAD
14003-402 76326-572 14003-398 14218-550 14003-396
Water, STERI-WATER® USP, purified, sterile
Water

Sterile water for chemical formulation, cleaning, and rinsing.


Filtered at 0.2 µm. Gamma-irradiated, double-bagged, and delivered with lot-specific sterile documentation.


Caution: Not for human or animal injection, diagnostic, or therapeutic use.

Formula: H₂O
MW: 18.02 g/mol
Boiling Pt: 100 °C (1013 hPa)
Melting Pt: 0 °C
MDL Number: MFCD00011332
CAS Number: 7732-18-5
Merck Index: 13,10098

Order Now

Specification Test Results

TOC <8.0 mg/L
Conductivity ≤5 µS/cm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR