STERI-PEROX, Veltek Associates
Supplier: Veltek Associates
SPER-16Z-6% SPER-01-6%
14003-384CS 1079.39 CAD
14003-384 75795-390
STERI-PEROX, Veltek Associates
Cleaning Agents Detergents and Surface Cleansers
Sterile, ready-to-use hydrogen peroxide solution for pharmaceutical and biotechnology operations. Formulated with USP Water for Injection
  • Filled in an ISO 5 (Grade A/B, Former Class 100)
  • Prefiltered to 0,2 um
  • Completely traceable and validated for sterility and shelf life
  • Avaliable in 3% and 6% concentrations
  • 16 oz and 32 oz containers come with sterile spray attachments
  • Low residue content
STERI-PEROX reduces exposure concerns for VOC’s in cleanroom operations, leaves a low remaining residue, and is designed for most washable, non-porous, hard, inanimate environmental surfaces.
Certifications: Delivered with lot specific Certificate of Analysis and Certificate of Sterility.
Packaging: The products are packaged to comply with the requirements for transportation into a classified area. Each container is double bagged packaged sterile.