STER-AHOL® WFI Formula, Veltek Associates
Supplier: Veltek Associates
DSTER-WFI-B-70 DSTER-WFI-SP-70 DSTER-WFI-70-5G
14003-354CS 1306.12 CAD
14003-354 14003-356 75795-620
STER-AHOL® WFI Formula, Veltek Associates
Cleaning Agents Disinfectants
Denatured ethanol formulated to 70% with a small percentage of methyl alcohol and 30% USP Water for Injection which can be used for the disinfection and decontamination of cleanrooms, controlled areas, hard non-porous environmental surfaces, and many other applications that require the use of a sterile alcohol solution.
  • Filled in ISO 5 (Grade A/B, Former Class 100)
  • Prefiltered to 0.2 um
  • Completely traceable and validated for sterility and shelf life
  • Gamma irradiated to a sterility assurance level of 10^-6.
  • Assayed according to current USP compendium