HYPO-CHLOR® Cleaning Agent, Veltek Associates
Supplier: Veltek
HYPO-CHLOR®
SHC-13Z-5.25 SHC-16Z-0.52 SHC-02-0.25 SHC-02-5.25 SHC-16Z-0.25 SHC-02-0.52 SHC-01-0.52 SHC-01-5.25 SHC-16Z-5.25
75795-370CS 609.36 CAD
14003-328 14003-336 14003-324 14218-554 75795-370 14003-334 14218-556 14003-333 14218-552
HYPO-CHLOR® Cleaning Agent, Veltek Associates
Cleaning Agents Detergents and Surface Cleansers
HYPO-CHLOR® is an effective one step, ready-to-use product that is available in premixed concentrations of 5.25%, 0.52%, and 0.25%.
  • Filled in ISO 5 (Grade A/B, Former Class 100)
  • Made with USP Water for Injection
  • Filtered at 0.2 microns
  • Sterility tested according to current USP compendium
  • Assayed according to current USP compendium
  • Completely traceable from start to finish
  • Completely validated for sterility and shelf life
Sodium hypochlorite solutions, formulated with WFI, used to clean cleanrooms and controlled areas including those used in health care institutions, biopharmaceutical, medical device, and diagnostic manufacturing facilities. HYPO-CHLOR® Products were developed for use in cleaning rotation cycles for maintaining a clean environment
Sterile sodium hypochlorite solutions are filtered at 0.2µm, gamma-irradiated, double-bagged, and delivered with lot-specific sterile documentation
Product uses: Use on hard, non-porous, and inanimate surfaces, on materials such as plastic, glass, vinyl, glazed porcelain, and stainless steel. Can also be used on machinery, tools, tables, counters, laminar flow benches, floors, walls and carts.
Delivery information: The 473 ml (16 oz.) sizes come with sterile spray attachments.