Sterile Nitrile Gloves, Hand-Specific, Kimtech™ G3 Sterling™ Sterile, Kimberly-Clark
Supplier: Microflex
11821 11824 11826 11827 11828 11822 11823 11825
CA89135-370CS 744.48 CAD
CA89135-370 CA89135-376 CA89135-380 CA89135-382 CA89135-384 CA89135-372 CA89135-374 CA89135-378
Sterile Nitrile Gloves, Hand-Specific, Kimtech™ G3 Sterling™ Sterile, Kimberly-Clark
Gloves
Powder-free, latex-free sterile gloves provide the protection of nitrile and sensitivity comparable to natural rubber latex.
  • Hand-specific
  • Beaded cuff
  • Textured fingertips
Static-dissipative in use. Recommended for ISO Class 4 (FED-STD-209E Class 1/M1.5) or less critical cleanroom environments. Walleted and pouched in polyethylene for cleanroom use and aseptic donning.
Certifications: PPE Category III product. Tested in accordance with EN Norms. Meets or exceeds AQL level of 1.5 for pinholes.
Ordering information: Each pair is double-packaged in a wallet within a pouch. Packaged in a Class 100 cleanroom.