VWR® Reclosable Bags
Supplier: VWR International
89071-798 89071-812 89071-814 89071-822 89071-824 89071-834 89071-846 89071-848 89071-850 89073-046 89073-056 89073-060 89073-062 89073-066 89073-068 89073-072 89073-074 89073-082 89073-084 89073-086 89073-088 89073-090 89073-092 89073-096 89073-100 89073-102 89073-104 89073-106 89073-108 89073-114 89073-116 89073-118 89073-122 89073-128 89073-132 89073-134
89071-798CS 875.73 CAD
89071-798 89071-812 89071-814 89071-822 89071-824 89071-834 89071-846 89071-848 89071-850 89073-046 89073-056 89073-060 89073-062 89073-066 89073-068 89073-072 89073-074 89073-082 89073-084 89073-086 89073-088 89073-090 89073-092 89073-096 89073-100 89073-102 89073-104 89073-106 89073-108 89073-114 89073-116 89073-118 89073-122 89073-128 89073-132 89073-134
VWR® Reclosable Bags
Bags Storage Bags
Reclosable bags are full gauge and offer exceptional clarity and quality.
  • LDPE resin fabrication
  • Reclosable design
  • Reinforced zipper locks in a smooth motion
Red line bags have a red line along the top of the bag and reinforced side-welded zipper to lock in a single smooth motion. Made of 100% virgin Low Density Polyethylene (LDPE) resin.
Certifications: Meet FDA/USDA specifications.