VWR® Tubing, PE
Supplier: VWR International
89071-044 89071-052 89071-038
89071-044CS 451.47 CAD
89071-044 89071-052 89071-038
VWR® Tubing, PE
Tubing Transfer Tubing
Polyethylene tubing is engineered to provide over three times the impact and twice the tensile strength of traditional low-density polyethylene.
  • Clear polyethylene construction
  • Heat sealable
  • Superior tensile strength to traditional LDPE
Clear polyethylene tubing is heat sealable. Wound on 7.6 cm (3") core. Core plug has a 2.5 cm (1") diameter hole. Manufactured with 100% renewable energy.
Order Now
SPECIFICATIONS
Environmentally Preferable