MicroSeal® VP Sealed Edge Polyester Knit Wipers, Berkshire
Supplier: Berkshire
MSVP.0909.8 MSVP1212B.14
94000-970CS 1520.4 CAD
94000-970 94000-972 94000-976
MicroSeal® VP Sealed Edge Polyester Knit Wipers, Berkshire
Wipes Cleanroom Wipes Dry Cleanroom Wipes
Wipers are made from 100% continuous filament knit polyester.
  • Lightweight, tight-knit construction resists abrasion, minimizes picking and snagging while cleaning rough surfaces
  • Cleanroom laundered wipers have ultrasonically sealed edges
Ideal for ISO Class 3 (FED-STD-209E Class 1/M1.5) and less critical environments. Works well with isopropyl alcohol, acetone, and other solvents. Suitable for cleaning areas with oxidation deposits, photolithography process areas, flat panels, and optics. Steam autoclavable for aseptic environments. Laundered andMore Product Information