Puritan® Tapered Mini Cotton Tipped Applicator, Wood Handle, Puritan Medical Products
Supplier: Puritan Medical Products
826-WC 25-8265WC 826-WC 25-8265WC 823-WC
CA10805-130CS 894 CAD
CA10805-130 CA89133-754 10805-130 89133-754 82004-051
Puritan® Tapered Mini Cotton Tipped Applicator, Wood Handle, Puritan Medical Products
Swabs Swabs for Clinical Use
This absorbent cotton tipped applicator is produced with high quality USP grade cotton, which is carded and coiled onsite to maintain consistency
It is bonded securely to a sturdy wood handle by an aqueous-based adhesive. The tapered mini cotton tip is ideal for hard-to-reach or small areas. Available nonsterile or sterile.