VWR® Wipers, Polyester
Supplier: VWR International
414004-517 414004-518
414004-517CS 1024.19 CAD
414004-517 414004-518
VWR® Wipers, Polyester
Wipes Cleanroom Wipes Dry Cleanroom Wipes
Durable wipers resist fiber release and abrasion under rigorous use, even when wiping rough surfaces.
  • 100% virgin, interlock knit polyester
  • Continuous filament
  • No-run material
  • Laser cut
  • Sealed edges
Laser-cut wipers with sealed edges limit particles and fibers to an absolute minimum, ensuring excellent performance in environments that demand low levels of contaminants.
Made from 100% virgin-grade, continuous-filament polyester in a double-knit, no-run interlock pattern to provide maximum cleanliness and absorbency. Designed for cleaning surfacesMore Product Information