VWR® Cleanroom Wipes, Nonwoven
Supplier: VWR International
414004-514 414004-515
414004-514CS 1420.77 CAD
414004-514 414004-515
VWR® Cleanroom Wipes, Nonwoven
Wipes Cleanroom Wipes Dry Cleanroom Wipes
These wipes are made from 55% polyester and 45% cellulose by hydroentanglement technology.
  • Manufactured and packed in cleanroom environment
  • Free from silicon, amides, and DOP contamination
  • Low lint and satisfactory softness to the touch
  • High absorbency
  • Tensile strength
  • Soft edges
These soft, economical wipers are designed for cleaning surfaces and controlling spills in ISO Class 6 (FED-STD-209E Class 1000/M4.5) and less critical environments.
Certifications: Wipers comply with RoHS 2002/95/EC.
Ordering information: Wipers are vacuum-packed in solvent-safe, cleanroom-compatible polyethylene with a label on each inner bag.