GammaGuard CE™ Frocks, Sterile, International ENVIROGUARD™
Supplier: International Enviroguard
CE11026CIS-M CE11026CIS-L CE11026CIS-XL CE11026CIS-2XL CE11026CIS-3XL CE11026CIS-4XL
89127-958CS 1027.35 CAD
89127-958 89127-960 89127-962 89127-964 89127-966 89127-968
GammaGuard CE™ Frocks, Sterile, International ENVIROGUARD™
Disposable Garments
GammaGuard CE™ sterile cleanroom apparel offers a superior liquid barrier, breathability, low linting, and tear resistance
Frocks are white with no pockets and tunnelized wrists. Frocks are sterilized to a 10-6 Sterility Assurance Level (SAL).
Special fold design prevents contamination during gowning. Material contains no natural rubber latex. Recyclable.
Ordering information: Manufactured in an ISO-certified facility, clean processed, and individually packaged under cleanroom conditions.