GammaGuard CE™ Coveralls, Sterile, International ENVIROGUARD™
Supplier: International Enviroguard
CE11013CIS-M CE11013CIS-L CE11013CIS-XL CE11013CIS-2XL CE11013CIS-3XL CE11013CIS-4XL
89127-946CS 773.54 CAD
89127-946 89127-948 89127-950 89127-952 89127-954 89127-956
GammaGuard CE™ Coveralls, Sterile, International ENVIROGUARD™
Disposable Garments
GammaGuard CE™ sterile cleanroom apparel offers a superior liquid barrier, breathability, low linting, and tear resistance
Coveralls are white with tunnelized wrists and ankles. Coveralls are sterilized to a 10-6 Sterility Assurance Level (SAL).
Special fold design prevents contamination during gowning. Material contains no natural rubber latex. Recyclable.
Ordering information: Manufactured in an ISO-certified facility, clean processed, and individually packaged under cleanroom conditions.