MicroGuard CE™ Coveralls, International ENVIROGUARD™
Supplier: International Enviroguard
CE8013CI-M CE8013CI-L CE8013CI-XL CE8013CI-2XL CE8013CI-3XL CE8013CI-4XL CE8019CI-M CE8019CI-L CE8019CI-XL CE8019CI-2XL CE8019CI-3XL CE8019CI-4XL
89127-702CS 674.79 CAD
89127-702 89127-704 89127-706 89127-708 89127-710 89127-712 89127-726 89127-728 89127-730 89127-732 89127-734 89127-736
MicroGuard CE™ Coveralls, International ENVIROGUARD™
Disposable Garments
MicroGuard CE™ cleanroom apparel combines a soft, non-woven fabric with a microporous film, providing a superior liquid barrier and good particulate barrier that is more breathable and low linting
Coveralls are white with tunnelized wrists and ankles.
Tear-resistant. Recyclable.