NovaSorb™ PolyCellulose™ String Mops, Micronova
Supplier: Micronova
CRMS-LBIR CRMS-LB JZCRMS-WH
89040-116CS 643.11 CAD
89040-116 89040-114 89501-296
NovaSorb™ PolyCellulose™ String Mops, Micronova
Mops
A highly absorbent flat strand mop made from a polyester/cellulose disposable material that outperforms standard cotton and rayon mops. Designed to cover large surface areas quickly and efficiently.
  • Highly absorbent and lightweight
  • Economically designed for single use in controlled environments
  • Outperforms standard cotton and rayon mops
  • Non-woven blend of 55% polyester and 45% cellulose disposable material
  • Holds up to strong disinfectants and common cleaning solutions
  • Available gamma irradiated for aseptic environments
  • Autoclavable
The Micronova NovaSorb™ PolyCellulose™ String Mop is designed for use with the Micronova MicroMop™ series hardware: MH-1 or MHC-1 handles, or QDMH-1 frame with SSU series handles.
Ordering information: All components must be ordered separately. For additional handle lengths, see Universal Mop Handles from Micronova.