Bottle Brush, Micronova
Supplier: Micronova
BRWH-01 BRWH-66 BRWH-67 BRYE-01 BRDG-01 BRRE-01 BRDB-01 BRWH-01B
89108-046CS 899.04 CAD
89108-046 89108-055 89108-053 89108-051 89187-751 76472-546 89187-753 89108-049
Bottle Brush, Micronova
Brushes
Multipurpose brushes with rounded polyester bristles for versatile cleaning of bottles and various equipment in labs, cleanrooms and production facilities. Handles are polypropylene. Brushes are BPA free.
  • Multipurpose, color-coded plastic bottle brushes
  • Durable polyester bristles
  • Stiff bristles for aggressive cleaning of bottles and equipment
  • Soft bristles for delicate cleaning and smaller openings
  • 12 or 16-inch easy grip, plastic handles
  • Made from FDA approved, BPA free base resins
  • Available in multiple colors
The Micronova M-Zone™Bottle Brushes are available with either stiff or soft polyester bristles and 12 or 16-inch plastic easy grip handles. The stiff bristle brushes are perfect for aggressive cleaning, while the softer bristle brushes feature a smaller diameter and are ideal for more delicate cleaning of jars and bottles with smaller openings.