KleenGuard® G10 Gray Nitrile Gloves, Kimberly-Clark
Supplier: Microflex
97820 97823
CA94001-396CS 762.46 CAD
CA94001-396 CA94001-398 CA94001-400 CA94001-402
KleenGuard® G10 Gray Nitrile Gloves, Kimberly-Clark
Gloves
Powder-free, ambidextrous gloves are manufactured from a nitrile formulation that has the fit and feel of latex.
  • These durable gloves for enhanced grip and dexterity
  • It has features of beaded cuffs and textured fingertips
  • Non sterile
Provides reliable protection in industrial and laboratory applications. An ideal replacement for latex gloves.
Ordering information: Gloves are packaged 150 per dispenser box, 10 dispenser boxes per case. Shipping case and inner packaging are labeled with product name, size, and lot/batch number.