Water USP, not sterile

Supplier: Ricca Chemical

Synonyms: Hydrogen oxide

R9191000-55E R9191000-330E3
RCR9191000-55EEA 1301.92 CAD
RCR9191000-55E RC9191000330E3
Water USP, not sterile
Water

The bulk water for this product is manufactured by a continuous process that has been validated under cGMP to produce high purity Water meeting, as a minimum, the specifications below at time of packaging.
This water is not filled in a sterile environment or into sterile containers. This water is not sterile and should not be used for injection or parenteral solution preparation.


This water is specially purified by conventional water softening, prefiltration, activated carbon organic adsorption, reverse osmosis, mixed bed triple deionization, ultraviolet light irradiation, and 0.2 micron membrane filtration.

Formula: H₂O
Boiling Pt: 100 °C
Melting Pt: 0 °C
Density: 0.997 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
EINECS: 231-791-2
Merck Index: 13,10098

Order Now

Specification Test Results

Ammonia (NH₃) max 0.3 ppm
Ammonium (NH₄) max 0.2 ppm
Appearance Colorless liquid
Calcium (Ca) max 0.5 ppm
Carbon Dioxide (CO₂) To Pass Test
Chloride (Cl) max 0.5 ppm
Conductivity at 25°C (Method: SQCP031) max 1 µS/cm
Magnesium (Mg) max 0.5 ppm
Organic Carbon (TOC) max 0.5 ppm
Oxidizable Substances (Permanganate Retention) To Pass Test
pH at 25°C (Method: SQCP027) 5.0-7.0
Sulfate (SO₄) max 0.5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR