Guanidinium hydrochloride ≥99.5%, white powder, GenAR® for biotechnology, Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
|
Warning
|
Management of Change (MOC) category = R
MACR0507-06
MACR0507-25
MACR0507-88
MACR0507-F7
MACR0507-F9
CAMK050704CS
1613.54
CAD
CAMK050704
CAMK050706
CAMK050725
CAMK050788
CAMK0507F7
CAMK0507F9
Guanidinium hydrochloride ≥99.5%, white powder, GenAR® for biotechnology, Macron Fine Chemicals™
Guanidine hydrochloride
White powder.
Formula:
NH₂C(=NH)NH₂·HCl Melting Pt: 185 °C Density: 1.354 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00013026 CAS Number: 50-01-1 EINECS: 200-002-3 Merck Index: 13,04578 |
|
Specification Test Results
GMP Manufactured Product | |
CAUTION: For Manufacturing, processing or repackaging | |
Assay (NH₂C(:NH)NH₂·HCl) | 99.5 - 101.0 % |
Acidity (as HCl) | ≤0.01 % |
Loss on Drying at 105°C | ≤0.5 % |
Residue after Ignition | ≤0.050 % |
Absorbance of a 6M Aqueous Solution - 230 nm | ≤0.200 |
Absorbance of a 6M Aqueous Solution - 260 nm | ≤0.030 |
Absorbance of a 6M Aqueous Solution - 275 nm | ≤0.030 |
RNase Activity (None Detected) | Passes Test |
DNase Activity (None Detected) | Passes Test |
Protease Activity (None Detected) | Passes Test |
Nitrate (NO₃) | ≤0.005 % |
Sulfate (SO₄) | ≤0.005 % |
Arsenic and Antimony (as As) | ≤0.500 ppm |
Trace Impurities - Copper (Cu) | ≤0.200 ppm |
Cyanide (CN) | ≤0.1 ppm |
Trace Impurities - Iron (Fe) | ≤2.0 ppm |
Trace Impurities - Lead (Pb) | ≤0.500 ppm |
Particulate Matter | Passes Test |
MANUFACTURED EXCLUSIVELY FOR AVANTOR PERFORMANCE MATERIALS, LLC., BY | |
BIOSPECTRA, INC., STROUDSBURG, PA |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...