Acetone ≥99.5%, Semi Grade for the electronics industry

Supplier: BDH

Synonyms: 2-PropanoneDMKPropanoneDimethyl ketone

10702S1G42BD
BDH2025-1GLPCS 525.42 CAD
BDH2025-1GLP
Acetone ≥99.5%, Semi Grade for the electronics industry
Acetone
Formula: CH₃COCH₃
MW: 58.08 g/mol
Boiling Pt: 56.2 °C (1013 hPa)
Melting Pt: –95.4 °C
Density: 0.792 g/cm³ (20 °C)
Flash Pt: <–20 °C (closed cup)
MDL Number: MFCD00008765
CAS Number: 67-64-1
UN: 1090
ADR: 3,II
Merck Index: 13,00067
https://ca.vwr.com/stibo/web/std.lang.all/06/07/720607.jpg

Order Now

Specification Test Results

Assay ([CH3]2CO) 99.5% Min.
Color (APHA) 10 APHA Max.
Acidity 0.3 μeq/g Max.
Alkalinity 0.5 μeq/g Max.
Residue after Evaporation 5 ppm Max.
Water (H2O) 0.5% Max.
Solubility in Water To Pass Test
Chloride (Cl) 200 ppb Max.
Phosphate (PO4) 100 ppb Max.
Arsenic and Antimony 100 ppb Max.
Al Impurities 100 ppb Max.
B Impurities 100 ppb Max.
Ca Impurities 100 ppb Max.
Cr Impurities 100 ppb Max.
Cu Impurities 100 ppb Max.
Au Impurities 100 ppb Max.
Fe Impurities 100 ppb Max.
Mg Impurities 100 ppb Max.
Mn Impurities 100 ppb Max.
Ni Impurities 100 ppb Max.
K Impurities 100 ppb Max.
Na Impurities 100 ppb Max.
Pb Impurities 100 ppb Max.
Sn Impurities 100 ppb Max.
Ti Impurities 100 ppb Max.
Zn Impurities 100 ppb Max.
Particles ≥1.0 μm 10/mL Max.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR